Få mere at vide
rac 1-Linoleoyl-2-linolenoyl-3-chloropropanediol, TRC
Artikelnummer. 30134121
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134121 10 mg
30008058 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134121 Leverandør Toronto Research Chemicals Leverandørnr. TRCL46808010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1612870-93-5
Molekylær formel C39H65ClO4
Molekylvægt (g/mol) 633.38
Synonym Linoleic-linolenic MCPD;
SMIL CCCCC/C=C\C/C=C\CCCCCCCC(=O)OCC(CCl)OC(=O)CCCCCCC/C=C\C/C=C\C/C=C\CC
Tekniske data
Kemisk navn eller materiale rac 1-Linoleoyl-2-linolenoyl-3-chloropropanediol
Mængde 10 mg
InChI formel InChI=1S/C39H65ClO4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h6,8,11-14,17-20,37H,3-5,7,9-10,15-16,21-36H2,1-2H3/b8-6-,13-11-,14-12-,19-17-,20-18-
Formel vægt 632.4571