Få mere at vide
rac 1-Linolenoyl-3-chloropropanediol-d5, TRC
Artikelnummer. 30478776
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30478776 25 mg -
30470891 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30478776 Leverandør Toronto Research Chemicals Leverandørnr. TRCL46770225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2734920-69-3
Molekylær formel C21H30D5ClO3
Molekylvægt (g/mol) 375.98
Synonym (Z,Z,Z)-9,12,15-Octadecatrienoic Acid 3-Chloro-2-hydroxypropyl-d5 Ester, 3-Chloro-1,2-propanediol-d5 1-Linolenate
IUPAC navn 3-chloro-2-hydroxypropyl-1,1,2,3,3-d5 (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
SMIL OC(C([2H])([2H])Cl)([2H])C([2H])([2H])OC(CCCCCCC/C=C\C/C=C\C/C=C\CC)=O
Tekniske data
Kemisk navn eller materiale rac 1-Linolenoyl-3-chloropropanediol-d5
Mængde 25 mg
InChI formel InChI=1S/C21H35ClO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h3-4,6-7,9-10,20,23H,2,5,8,11-19H2,1H3/b4-3-,7-6-,10-9-/i18D2,19D2,20D
Formel vægt 375.26