Få mere at vide
rac 1-Lauroyl-3-chloropropanediol, TRC
Artikelnummer. 30483012
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483012 25 mg
30480673 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30483012 Leverandør Toronto Research Chemicals Leverandørnr. TRCL18475025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 20542-96-5
Molekylær formel C15 H29 Cl O3
Molekylvægt (g/mol) 292.84
IUPAC navn (3-chloro-2-hydroxypropyl) dodecanoate
SMIL CCCCCCCCCCCC(=O)OCC(O)CCl
Tekniske data
Kemisk navn eller materiale 1-Lauroyl-3-chloropropanediol
Mængde 25 mg
InChI formel InChI=1S/C15H29ClO3/c1-2-3-4-5-6-7-8-9-10-11-15(18)19-13-14(17)12-16/h14,17H,2-13H2,1H3
Formel vægt 292.1805