Få mere at vide
rac 1-Chloro-3-[4-(2-methoxyethyl)phenoxy]-2-propanol, TRC
Artikelnummer. 30359743
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359743 25 mg
30351707 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30359743 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36798025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Chloro-3-[4-(2-methoxyethyl)-phenoxy]propan-2-ol
CAS 56718-76-4
Molekylær formel C12 H17 Cl O3
Mængde 25 mg
Synonym 1-Chloro-3-[4-(2-methoxyethyl)-phenoxy]propan-2-ol
InChI formel InChI=1S/C12H17ClO3/c1-15-7-6-10-2-4-12(5-3-10)16-9-11(14)8-13/h2-5,11,14H,6-9H2,1H3
SMIL COCCc1ccc(OCC(O)CCl)cc1
IUPAC navn 1-chloro-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol
Molekylvægt (g/mol) 244.71
Formel vægt 244.0866