Få mere at vide
rac 1,2-Bis-palmitoyl-3-chloropropanediol, TRC
Artikelnummer. 17928604
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17928604 10 mg
17908434 50 mg
17948704 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17928604 Leverandør Toronto Research Chemicals Leverandørnr. TRCB51520010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 51930-97-3
Molekylær formel C35 H67 Cl O4
Molekylvægt (g/mol) 587.36
Synonym Hexadecanoic acid, 1,1'-[1-(chloromethyl)-1,2-ethanediyl] ester, Hexadecanoic acid, 1-(chloromethyl)-1,2-ethanediyl ester (9CI), 3-Chloropropane-1,2-diol dipalmitate, Rac-1,2-bis(palmitoyl)-3-chloropropane-1,2-diol, rac 1,2-Bis-palmitoyl-3-chloropropanediol
IUPAC navn (3-chloro-2-hexadecanoyloxypropyl) hexadecanoate
SMIL CCCCCCCCCCCCCCCC(=O)OCC(CCl)OC(=O)CCCCCCCCCCCCCCC
Tekniske data
Kemisk navn eller materiale rac-1,2-Bis-palmitol-3-chloropropanediol
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C35H67ClO4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(37)39-32-33(31-36)40-35(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33H,3-32H2,1-2H3
Formel vægt 586.4728
Procent renhed >95%