Få mere at vide
Rabeprazole Sulfide N-Oxide, TRC
Artikelnummer. 17954295
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17954295 50 mg
17940455 25 mg
17934375 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17954295 Leverandør Toronto Research Chemicals Leverandørnr. TRCR07053550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 924663-40-1
Molekylær formel C18 H21 N3 O3 S
Molekylvægt (g/mol) 359.44
Synonym 2-[[[4-(3-Methoxypropoxy)-3-methyl-1-oxido-2-pyridinyl]methyl]thio]-1H-benzimidazole, Rabeprazole Sulfide N-Oxide
IUPAC navn 2-(1H-benzimidazol-2-ylsulfanylmethyl)-4-(3-methoxypropoxy)-3-methylpyridine 1-oxide
SMIL COCCCOc1ccn(=O)c(CSc2nc3ccccc3[nH]2)c1C
Tekniske data
Kemisk navn eller materiale Rabeprazole Sulfide N-Oxide
Mængde 50 mg
InChI formel InChI=1S/C18H21N3O3S/c1-13-16(21(22)9-8-17(13)24-11-5-10-23-2)12-25-18-19-14-6-3-4-7-15(14)20-18/h3-4,6-9H,5,10-12H2,1-2H3,(H,19,20)
Formel vægt 359.1304