Få mere at vide
Rabbit Pure-Blot anti-Mouse IgG Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23974
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG Polyclonal antibody specifically detects IgG in Mouse samples. It is validated for ELISA
Tekniske data
| IgG | |
| Polyclonal | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.1 mg/mL Bovine Serum Albumin (BSA) - IgG and Protease free, 50% (v/v) Glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Mouse | |
| Purified |
| ELISA | |
| Unconjugated | |
| Immunoglobulin G, ImmunoglobulinG | |
| highly purified mouse IgG gamma 1, gamma 2a, gamma 2b and gamma 3 proteins | |
| 1 mg | |
| Adaptive Immunity, Epitope Tags, Immunology | |
| 3500 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |