Få mere at vide
USP20 Rabbit anti-Bovine, Chicken, Dog, Horse, Human, Monkey, Pig, Rabbit, Polyclonal Antibody, Abnova™
Tekniske data
Tekniske data
| Antigen | USP20 |
| Applikationer | Immunohistochemistry (PFA fixed) |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | Rabbit polyclonal antibody raised against synthetic peptide of USP20. |
| Fortynding | Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (0.09% sodium azide) |
| Gene | USP20 |
| Gene Alias | KIAA1003/LSFR3A/VDU2 |
| Gensymboler | USP20 |
| Vis mere |
For Research Use Only