missing translation for 'onlineSavingsMsg'
Få mere at vide

SURF4 Rabbit anti-Bovine, Chicken, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Artikelnummer. 16087907
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Pakningsstørrelse:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16087907 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16087907 Leverandør Abnova Leverandørnr. PAB28180.50ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit polyclonal antibody raised against synthetic peptide of SURF4.

This gene is located in the surfeit gene cluster, which is comprised of very tightly linked housekeeping genes that do not share sequence similarity. The encoded protein is a conserved integral membrane protein containing multiple putative transmembrane regions. In eukaryotic cells, protein transport between the endoplasmic reticulum and Golgi compartments is mediated in part by nonclathrin-coated vesicular coat proteins (COPs). The specific function of this protein has not been determined but its yeast homolog is directly required for packaging glycosylated pro-alpha-factor into COPII vesicles. This gene uses multiple polyadenylation sites, resulting in transcript length variation. The existence of alternatively spliced transcript variants has been suggested, but their validity has not been determined. [provided by RefSeq

Tekniske data

Antigen SURF4
Applikationer Immunohistochemistry (PFA fixed)
Klassifikation Polyclonal
Konjugeret Unconjugated
Oversigt Rabbit polyclonal antibody raised against synthetic peptide of SURF4.
Fortynding Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15 ug/ml)
Formulering In PBS (0.09% sodium azide)
Gene SURF4
Gene Alias ERV29/FLJ22993/MGC102753
Gensymboler SURF4
Værtsarter Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acids at N-terminus region of human SURF4.
Oprensningsmetode Immunoaffinity chromatography
Mængde 50 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 6836
Testspecificitet BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Målarter Bovine, Chicken, Human, Mouse, Rat
Indhold og opbevaring Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Vis mere Vis mindre

For Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.