missing translation for 'onlineSavingsMsg'
Få mere at vide

RAD51 Rabbit anti-Chicken, Human, Mouse, Polyclonal Antibody, Abnova™

Artikelnummer. 16101930
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μL
Pakningsstørrelse:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16101930 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16101930 Leverandør Abnova Leverandørnr. PAB1184.50uL

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit polyclonal antibody raised against full-length recombinant RAD51.

The protein encoded by this gene is a member of the RAD51 protein family. RAD51 family members are highly similar to bacterial RecA and Saccharomyces cerevisiae Rad51, and are known to be involved in the homologous recombination and repair of DNA. This protein can interact with the ssDNA-binding protein RPA and RAD52, and it is thought to play roles in homologous pairing and strand transfer of DNA. This protein is also found to interact with BRCA1 and BRCA2, which may be important for the cellular response to DNA damage. BRCA2 is shown to regulate both the intracellular localization and DNA-binding ability of this protein. Loss of these controls following BRCA2 inactivation may be a key event leading to genomic instability and tumorigenesis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Tekniske data

Antigen RAD51
Applikationer Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Klassifikation Polyclonal
Konjugeret Unconjugated
Oversigt Rabbit polyclonal antibody raised against full length recombinant RAD51.
Fortynding Western Blot (1:2000-1:10000) The optimal working dilution should be determined by the end user.
Formulering In buffer containing 0.09% sodium azide
Gene RAD51
Gene Alias BRCC5/HRAD51/HsRad51/HsT16930/RAD51A/RECA
Gensymboler RAD51
Værtsarter Rabbit
Immunogen Recombinant protein corresponding to full length human RAD51.
Mængde 50 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 5888
Målarter Chicken, Human, Mouse
Indhold og opbevaring Store the kit at -20°C.
Form Liquid
Vis mere Vis mindre

For Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.