missing translation for 'onlineSavingsMsg'
Få mere at vide

PYGB Rabbit anti-Bovine, Chicken, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Zebra fish, Polyclonal Antibody, Abnova™

Artikelnummer. 16001957
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Pakningsstørrelse:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16001957 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16001957 Leverandør Abnova Leverandørnr. PAB16299.50ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit polyclonal antibody raised against synthetic peptide of PYGB.

The protein encoded by this gene is a glycogen phosphorylase found predominantly in the brain. The encoded protein forms homodimers which can associate into homotetramers, the enzymatically active form of glycogen phosphorylase. The activity of this enzyme is positively regulated by AMP and negatively regulated by ATP, ADP, and glucose-6-phosphate. This enzyme catalyzes the rate-determining step in glycogen degradation. [provided by RefSeq

Tekniske data

Antigen PYGB
Applikationer Immunohistochemistry (PFA fixed)
Klassifikation Polyclonal
Konjugeret Unconjugated
Oversigt Rabbit polyclonal antibody raised against synthetic peptide of PYGB.
Fortynding Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulering In PBS (0.09% sodium azide)
Gene PYGB
Gene Alias MGC9213
Gensymboler PYGB
Værtsarter Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human PYGB.
Oprensningsmetode Immunoaffinity purification
Mængde 50 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 5834
Testspecificitet Internal domain of human.
Målarter Bovine, Chicken, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Indhold og opbevaring Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Vis mere Vis mindre

For Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.