missing translation for 'onlineSavingsMsg'
Få mere at vide

PTPRE Rabbit anti-Bats, Bovine, Chicken, Dog, Hamster, Horse, Human, Monkey, Polyclonal Antibody, Abnova™

Artikelnummer. 16067937
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Pakningsstørrelse:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16067937 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16067937 Leverandør Abnova Leverandørnr. PAB28343.50ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit polyconal antibody raised against synthetic peptide of PTPRE.

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. Two alternatively spliced transcript variants of this gene have been reported, one of which encodes a receptor-type PTP that possesses a short extracellular domain, a single transmembrane region, and two tandem intracytoplasmic catalytic domains; Another one encodes a PTP that contains a distinct hydrophilic N-terminus, and thus represents a nonreceptor-type isoform of this PTP. Studies of the similar gene in mice suggested the regulatory roles of this PTP in RAS related signal transduction pathways, cytokines induced SATA signaling, as well as the activation of voltage-gated K+ channels. [provided by RefSeq]

Tekniske data

Antigen PTPRE
Applikationer Immunohistochemistry (PFA fixed)
Klassifikation Polyclonal
Konjugeret Unconjugated
Oversigt Rabbit polyconal antibody raised against synthetic peptide of PTPRE.
Fortynding Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-6 ug/ml) The optimal working dilution should be determined by the end user.
Formulering In PBS (0.09% sodium azide)
Gene PTPRE
Gene Alias DKFZp313F1310/FLJ57799/FLJ58245/HPTPE/PTPE/R-PTP-EPSILON
Gensymboler PTPRE
Værtsarter Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acids at cytoplasmic domain of human PTPRE.
Oprensningsmetode Immunoaffinity chromatography
Mængde 50 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 5791
Testspecificitet BLAST analysis of the peptide immunogen showed no homology with other human proteins, except PTPRA (67%).
Målarter Bat, Bovine, Canine, Chicken, Equine, Hamster, Human, Monkey
Indhold og opbevaring Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Vis mere Vis mindre

For Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.