missing translation for 'onlineSavingsMsg'
Få mere at vide

PARK7 Rabbit anti-Bovine, Chicken, Hamster, Human, Mouse, Rat, Zebra fish, Polyclonal Antibody, Abnova™

Artikelnummer. 16039676
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
Pakningsstørrelse:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16039676 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16039676 Leverandør Abnova Leverandørnr. PAB12080.100uL

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit polyclonal antibody raised against synthetic peptide of PARK7.

The product of this gene belongs to the peptidase C56 family of proteins. It acts as a positive regulator of androgen receptor-dependent transcription. It may also function as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. Defects in this gene are the cause of autosomal recessive early-onset Parkinson disease 7. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq

Tekniske data

Antigen PARK7
Applikationer Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Klassifikation Polyclonal
Konjugeret Unconjugated
Oversigt Rabbit polyclonal antibody raised against synthetic peptide of PARK7.
Fortynding Immunocytochemistry (1:500) Immunoprecipitation (1:200) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulering No additive
Gene PARK7
Gene Alias DJ-1/DJ1/FLJ27376/FLJ34360/FLJ92274
Gensymboler PARK7
Værtsarter Rabbit
Immunogen A synthetic peptide corresponding to amino acids 150-189 of human PARK7.
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 11315
Testspecificitet This antibody is specific to DJ-1.
Målarter Bovine, Chicken, Hamster, Human, Mouse, Rat, Zebrafish
Indhold og opbevaring Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Vis mere Vis mindre

For Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.