missing translation for 'onlineSavingsMsg'
Få mere at vide

NTRK1 (phospho Y757), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Artikelnummer. 16294210
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
Pakningsstørrelse:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16294210 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16294210 Leverandør Abnova Leverandørnr. PAB29221.100uL

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human NTRK1.

This gene encodes a member of the neurotrophic tyrosine kinase receptor (NTKR) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of this kinase leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in this gene have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer. Alternate transcriptional splice variants of this gene have been found, but only three have been characterized to date. [provided by RefSeq

Tekniske data

Antigen NTRK1
Applikationer Immunohistochemistry (PFA fixed)
Klassifikation Polyclonal
Konjugeret Unconjugated
Oversigt Rabbit polyclonal antibody raised against synthetic phosphopeptide of human NTRK1.
Fortynding Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulering In PBS, pH 7.4 (without Mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide)
Gene NTRK1
Gene Alias DKFZp781I14186/MTC/TRK/TRK1/TRKA/p140-TrkA
Gensymboler NTRK1
Værtsarter Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y757 of human NTRK1.
Oprensningsmetode Affinity purification
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 4914
Testspecificitet This antibody detects endogenous levels of NTRK1 only when phosphorylated at tyrosine 757.
Målarter Human, Mouse
Indhold og opbevaring Store at -20°C.
Avoid repeated freezing and thawing.
Form Liquid
Vis mere Vis mindre

For Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.