missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
NOC2L, Rabbit, Polyclonal Antibody, Abnova™
Beskrivelse
Histone modification by histone acetyltransferases (HAT) and histone deacetylases (HDAC) can control major aspects of transcriptional regulation. NOC2L represents a novel HDAC-independent inhibitor of histone acetyltransferase (INHAT) (Hublitz et al., 2005 [PubMed 16322561]).[supplied by OMIM
Sequence: LTVDEFLASGFDSESESESENSPQAETREAREAARSPDKPGGSPSASRRKGRASEHKDQLSRLKDRDPEFYKFLQENDQSLLNFSDS
Tekniske data
Tekniske data
| Antigen | NOC2L |
| Applikationer | Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | Rabbit polyclonal antibody raised against recombinant NOC2L. |
| Fortynding | Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) |
| Gene | NOC2L |
| Gene Alias | DKFZp564C186/FLJ35172/NIR |
| Gensymboler | NOC2L |
| Vis mere |
For Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?