missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Rabbit anti-Mouse Lambda Light Chain Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72792-0.5mg
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Lambda Light Chain Polyclonal antibody specifically detects Lambda Light Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Tekniske data
| Lambda Light Chain | |
| Polyclonal | |
| Western Blot 1:1000 - 1:10000, ELISA 1:40000, Immunohistochemistry 1:500 - 1:3000 | |
| IGLC, immunoglobulin lambda constant group | |
| Mouse lambda light chain | |
| 0.5 mg | |
| Adaptive Immunity, Immunology | |
| 3537 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Mouse | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion