missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Rabbit anti-Mouse IgG F(ab')2 Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-73945-2mg
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Tekniske data
| IgG F(ab')2 | |
| Polyclonal | |
| Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| Mouse IgG F(ab')2 fragment | |
| 2 mg | |
| Adaptive Immunity, Epitope Tags, Immunology | |
| Mouse | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion