missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
LOC780933 (Trypsin (Bovine)), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native Trypsin (Bovine).
Brand: Abnova PAB28070.100ug
Denne vare kan ikke returneres.
Se returpolitik
Tekniske data
| LOC780933 (Trypsin (Bovine)) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native Trypsin (Bovine). | |
| Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| MGC157392 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Bovine | |
| Lyophilized |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:1000-1:4000) The optimal working dilution should be determined by the end user. | |
| LOC780933 | |
| LOC780933 | |
| Native purified bovine Trypsin from bovine pancreas. | |
| RUO | |
| 780933 | |
| Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion