Få mere at vide
Clostridium botulinum Type E Neurotoxin Complex, Rabbit, Polyclonal Antibody, Abnova™
Tekniske data
Tekniske data
| Antigen | Clostridium botulinum Type E Neurotoxin Complex |
| Applikationer | Dot Blot, ELISA, Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | Rabbit polyclonal antibody raised against Clostridium botulinum Type E Neurotoxin Complex. |
| Fortynding | The optimal working dilution should be determined by the end user. |
| Formulering | In PBS, pH 7.2 (20% glycerol) |
| Værtsarter | Rabbit |
| Immunogen | Clostridium botulinum type E Neurotoxin and complexing Proteins. |
| Mængde | 1 mg |
| Vis mere |
For Research Use Only