Få mere at vide
Rabbit anti-Hamster IgG (H+L) Secondary Antibody, DyLight 633, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76177
2265.25 DKK valid until 2025-12-16
BEDSTE tilbudspris! Brug kampagnekoden "24090" for at få din tilbudspris.
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Hamster samples. It is validated for Flow Cytometry, Immunomicroscopy.
Tekniske data
| IgG (H+L) | |
| Polyclonal | |
| DyLight 633 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. | |
| Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage. |
| Flow Cytometry, Immunomicroscopy | |
| LYOPH | |
| Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000 | |
| Purified Hamster IgG (H&L) | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| Based on IEP, this antibody reacts with heavy gamma chains on hamster IgG and light chains on all hamster immunoglobulins | |
| Hamster | |
| IgG |
Korrektion af produktindhold
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion