missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Rabbit anti-Golden Syrian Hamster IgG (H+L) Secondary Antibody, Texas Red, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-73364
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Golden Syrian Hamster samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay
Tekniske data
| IgG (H+L) | |
| Polyclonal | |
| Texas Red | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
| Hamster IgG whole molecule | |
| 1.5 mg | |
| Adaptive Immunity, Immunology | |
| Reconstitute with 1.0 mL deionized water (or equivalent). | |
| Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-Linked Immunosorbent Assay | |
| LYOPH | |
| Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Golden Syrian Hamster | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion