missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Rabbit anti-Goat IgG (H+L) Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
815.00 DKK - 1695.00 DKK
Tekniske data
| Antigen | IgG (H+L) |
|---|---|
| Fortynding | Western Blot 1:100-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunoassay |
| Applikationer | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoassay |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
18543680
|
Novus Biologicals
NBP1-74825 |
2 mg |
1695.00 DKK
2mg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
18592572
|
Novus Biologicals
NBP1-74825-0.5mg |
0.5 mg |
815.00 DKK
0.50mg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, ImmunoassayTekniske data
| IgG (H+L) | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoassay | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Immunology | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunoassay | |
| Polyclonal | |
| Purified | |
| RUO | |
| Goat | |
| This Rabbit anti-Goat IgG (H+L) Secondary Antibody was developed against purified goat IgG (H&L). | |
| Secondary | |
| Store at 4°C. Do not freeze. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel