Få mere at vide
FNTA Rabbit anti-Chicken, Chimpanzee, Dog, Human, Rat, Polyclonal Antibody, Abnova™
Beskrivelse
Prenyltransferases attach either a farnesyl group or a geranylgeranyl group in thioether linkage to the cysteine residue of protein's with a C-terminal CAAX box. CAAX geranylgeranyltransferase and CAAX farnesyltransferase are heterodimers that share the same alpha subunit but have different beta subunits. This gene encodes the alpha subunit of these transferases. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Tekniske data
Tekniske data
| Antigen | FNTA |
| Applikationer | Immunohistochemistry, Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | Rabbit polyclonal antibody raised against partial recombinant FNTA. |
| Fortynding | The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (0.2% gelatin, 0.05% sodium azide) |
| Gene | FNTA |
| Genadgangsnr. | NP_002018 |
| Gene Alias | FPTA/MGC99680/PGGT1A/PTAR2 |
| Vis mere |
For Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.