missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Rabbit anti-Feline IgG F(ab')2 Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-73919
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Feline samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Tekniske data
| IgG F(ab')2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| Cat IgG F(ab')2 fragment | |
| 50 mg | |
| Adaptive Immunity, Epitope Tags, Immunology | |
| Reconstitute with 5.0 mL deionized water (or equivalent). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Feline | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion