Learn More
FAXC, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FAXC.
Brand: Abnova PAB23398.100uL
This item is not returnable.
View return policy
Description
Sequence: HFYWTLAYCQWVDNLNETRKMLSLSGGGPFSNLLRWVVCHITKGIVKREMHGHGIGRFSEEEIYMLMEKDMRSLAGLLGSpecifications
FAXC | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant FAXC. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
Q5TGI0 | |
FAXC | |
Recombinant protein corresponding to amino acids of human FAXC. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
FAXC | |
C6orf168/dJ273F20 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
84553 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Product Content Correction
Product Title
FAXC, Rabbit, Polyclonal Antibody, Abnova™ > 100μL; Unlabeled
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu