Få mere at vide
ELTD1 Rabbit anti-Chicken, Horse, Human, Mouse, Pig, Rabbit, Rat, Polyclonal Antibody, Abnova™
Tekniske data
Tekniske data
| Antigen | ELTD1 |
| Applikationer | Immunohistochemistry (PFA fixed) |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | Rabbit polyclonal antibody raised against synthetic peptide of ELTD1. |
| Fortynding | Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-11 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (0.09% sodium azide) |
| Gene | ELTD1 |
| Genadgangsnr. | Q9HBW9 |
| Gene Alias | ETL/KPG_003 |
| Vis mere |
For Research Use Only