missing translation for 'onlineSavingsMsg'
Få mere at vide

EGFR (phospho S1142), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Artikelnummer. 16070718
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
Pakningsstørrelse:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16070718 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16070718 Leverandør Abnova Leverandørnr. PAB7901.100uL

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit polyclonal antibody raised against synthetic phosphopeptide of EGFR.

The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. [provided by RefSeq

Tekniske data

Antigen EGFR
Applikationer ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Klassifikation Polyclonal
Konjugeret Unconjugated
Oversigt Rabbit polyclonal antibody raised against synthetic phosphopeptide of EGFR.
Fortynding ELISA (1:2000) Immunocytochemistry (1:200) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulering In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide)
Gene EGFR
Gene Alias ERBB/ERBB1/HER1/PIG61/mENA
Gensymboler EGFR
Værtsarter Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S1142 of human EGFR.
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 1956
Testspecificitet This human EGFR sequence has high homology with rat and mouse EGFR, and is not conserved in other ErbB family members.
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Vis mere Vis mindre

For Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.