missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CYP3A4/CYP3A5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CYP3A4/CYP3A5.
Brand: Abnova PAB24907.100uL
Denne vare kan ikke returneres.
Se returpolitik
Tekniske data
| CYP3A4/CYP3A5 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| CP33/CP34/CYP3A/CYP3A3/HLP/MGC126680/NF-25/P450C3/P450PCN1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1576/1577 | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of CYP3A4/CYP3A5. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| CYP3A4 | |
| A synthetic peptide corresponding to CYP3A4/CYP3A5. | |
| 100 μL | |
| Primary | |
| CYP3A4/CYP3A5 polyclonal antibody detects endogenous levels of CYP3A4/CYP3A5 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion