missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Rabbit anti-Chicken IgY (H+L) Secondary Antibody

Artikelnummer. 15353936
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2 mg
Pakningsstørrelse:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15353936 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15353936 Leverandør Invitrogen™ Leverandørnr. A16132

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Secondary Antibody

The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).

Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Chicken IgY (H+L)
Applikationer ELISA, Western Blot
Klassifikation Polyclonal
Koncentration >4.5 mg/mL
Konjugeret Unconjugated
Formulering PBS with 0.05% sodium azide; pH 7.2
Værtsarter Rabbit
Immunogen Chicken IgY (H+L).
Oprensningsmetode Affinity chromatography
Mængde 2 mg
Regulatorisk status RUO
Primær eller sekundær Secondary
Målarter Chicken
Indhold og opbevaring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttype Secondary Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.