missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Rabbit anti-Chicken IgY (H+L) Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
1210.00 DKK - 2320.00 DKK
Tekniske data
| Antigen | IgY (H+L) |
|---|---|
| Fortynding | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunodiffusion, Immunoassay |
| Applikationer | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Product Code | Brand | Mængde | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Mængde | Price | Quantity & Availability | |||||
|
18578592
|
Novus Biologicals
NBP1-74781 |
2 mg |
2320.00 DKK
2mg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
18557710
|
Novus Biologicals
NBP1-74781-0.5mg |
0.5 mg |
1210.00 DKK
0.50mg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Description
IgY (H+L) Polyclonal antibody specifically detects IgY (H+L) in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, ImmunoassaySpecifications
| IgY (H+L) | |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion | |
| Unconjugated | |
| Rabbit | |
| Chicken | |
| Purified Chicken IgG/Y (H&L) | |
| Secondary | |
| Store at 4°C. Do not freeze. |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunodiffusion, Immunoassay | |
| Polyclonal | |
| Purified | |
| RUO | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title