Få mere at vide
Invitrogen™ Rabbit anti-Chicken IgY (H+L) Secondary Antibody
Beskrivelse
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31104 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31104 reacts with the heavy chains of chicken IgY and with the light chains common to most chicken immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Note: IgY is the original designation for the IgG like protein found in both serum and egg yolk. Country of origin: USA.
Tekniske data
Tekniske data
| Antigen | Chicken IgY (H+L) |
| Applikationer | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Koncentration | 2.38 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with no preservative; pH 7.6 |
| Værtsarter | Rabbit |
| Oprensningsmetode | Antigen affinity chromatography |
| Mængde | 2 mg |
| Regulatorisk status | RUO |
| Vis mere |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.