Få mere at vide
CANX Rabbit anti-Birds, Bovine, Chicken, Dog, Frog, Fruit fly, Guinea pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Polyclonal Antibody, Abnova™
Tekniske data
Tekniske data
| Antigen | CANX |
| Applikationer | Immunohistochemistry, Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | Rabbit polyclonal antibody raised against synthetic peptide of CANX. |
| Fortynding | Western Blot (1:2000) The optimal working dilution should be determined by the end user. |
| Formulering | In antiserum |
| Gene | CANX |
| Gene Alias | PP90 |
| Gensymboler | CANX |
| Vis mere |
For Research Use Only