missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Rabbit anti-Canine IgG F(ab')2 Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-73923
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Tekniske data
| IgG F(ab')2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| Dog IgG F(ab')2 fragment | |
| 50 mg | |
| Adaptive Immunity, Epitope Tags, Immunology | |
| Reconstitute with 5.0 mL deionized water (or equivalent). | |
| Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Canine | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido