missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C12orf23, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant C12orf23.
Brand: Abnova PAB23927.100uL
Additional Details : Weight : 0.02000kg
Description
Sequence: QQEIPSYLNDEPPEGSMKDHPQQQPGMLSRSpecifications
C12orf23 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant C12orf23. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
Q8WUH6 | |
C12orf23 | |
Recombinant protein corresponding to amino acids of human C12orf23. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
C12orf23 | |
FLJ11721/FLJ13959/MGC17943 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
90488 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
For Research Use Only