missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADCY5/ADCY6, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ADCY5/ADCY6.
Brand: Abnova PAB17485.100ug
This item is not returnable.
View return policy
Specifications
ADCY5/ADCY6 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ADCY5/ADCY6. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
AC5 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total ADCY5/ADCY6 protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
O43306/O95622 | |
ADCY5 | |
A synthetic peptide corresponding to internal of human ADCY5/ADCY6. | |
RUO | |
111/112 | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ADCY5/ADCY6, Rabbit, Polyclonal Antibody, Abnova™ > 100μg; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction