Få mere at vide
Invitrogen™ RABAC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120688
Beskrivelse
Positive test controls include: 293T, U-251MG, SGC-7901, Mouse lung, Rat testis, Rat ovary. Immunogen sequence: MAAQKDQQKD AEAEGLSGTT LLPKLIPSGA GREWLERRRA TIRPWSTFVD QQRFSRPRNL GELCQRLVRN VEYYQSNYVF VFLGLILYCV VTSPMLLVAL AVFFGACYIL YLRTLESKLV LFGREVSPAH QYALAGGISF PFFWLAGAGS AVFWVLGATL VVIGSHAAFH QIEAVDGEEL QMEPV.
Tekniske data
| RABAC1 | |
| Polyclonal | |
| Unconjugated | |
| RABAC1 | |
| 2310040I06Rik; Gbpap1; GTP binding protein associated protein 1; Pra; Pra1; PRA1 domain family 1; PRA1 family protein 1; Praf1; prenylated Rab acceptor 1; prenylated Rab acceptor protein 1; Prenylin; Rab acceptor 1; Rab acceptor 1 (prenylated); RABAC1; YIP3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10567, 14470, 83583 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.69 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O35394, Q9UI14, Q9Z0S9 | |
| RABAC1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-185 of human RABAC1 (NP_006414.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.