missing translation for 'onlineSavingsMsg'
Få mere at vide

(R)-Viloxazine Hydrochloride, TRC

Artikelnummer. 30450620
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30450620 50 mg
30456923 10 mg
30451623 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30450620 Leverandør Toronto Research Chemicals Leverandørnr. TRCV31245050MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56287-63-9
Molekylær formel C13H19NO3 • HCl
Molekylvægt (g/mol) 237.293646
Synonym (R)-2-[(2-Ethoxyphenoxy)methyl]morpholine Hydrochloride, (R)-2-(2-Ethoxyphenoxymethyl)tetrahydro-1,4-oxazine Hydrochloride
IUPAC navn (2R)-2-[(2-ethoxyphenoxy)methyl]morpholine;hydrochloride
SMIL CCOC1=CC=CC=C1OC[C@H]2CNCCO2.Cl

Specifications

Kemisk navn eller materiale (R)-Viloxazine Hydrochloride
Mængde 50 mg
InChI formel InChI=1S/C13H19NO3.ClH/c1-2-15-12-5-3-4-6-13(12)17-10-11-9-14-7-8-16-11;/h3-6,11,14H,2,7-10H2,1H3;1H/t11-;/m1./s1
Formel vægt 273.1132
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.