Få mere at vide
(R)-Viloxazine-d5 Hydrochloride, TRC
Artikelnummer. 30451392
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30451392 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451392 Leverandør Toronto Research Chemicals Leverandørnr. TRCV31245210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-Viloxazine-d5 Hydrochloride
CAS 1246815-04-2
Molekylær formel C13H15D5ClNO3
Mængde 10 mg
Synonym (R)-2-[((2-Ethoxy-d5)phenoxy)methyl]morpholine Hydrochloride, (R)-2-[(2-Ethoxy-d5)phenoxymethyl]tetrahydro-1,4-oxazine Hydrochloride
InChI formel InChI=1S/C13H19NO3.ClH/c1-2-15-12-5-3-4-6-13(12)17-10-11-9-14-7-8-16-11;/h3-6,11,14H,2,7-10H2,1H3;1H/t11-;/m1./s1/i1D3,2D2;
SMIL Cl.[2H]C([2H])([2H])C([2H])([2H])Oc1ccccc1OC[C@H]2CNCCO2
IUPAC navn (2R)-2-[[2-(1,1,2,2,2-pentadeuterioethoxy)phenoxy]methyl]morpholine;hydrochloride
Molekylvægt (g/mol) 278.79
Formel vægt 278.1446