Få mere at vide
R-Venlafaxine, TRC
Artikelnummer. 30137697
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137697 50 mg
30005650 5 mg
30008248 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137697 Leverandør Toronto Research Chemicals Leverandørnr. TRCV12000350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 93413-46-8
Molekylær formel C17 H27 N O2
Molekylvægt (g/mol) 277.4
Synonym Cyclohexanol, 1-[(1R)-2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]-, Cyclohexanol, 1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]-, (R)-, 1-[(1R)-2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol, (R)-Venlafaxine
IUPAC navn 1-[(1R)-2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol
SMIL COc1ccc(cc1)[C@H](CN(C)C)C2(O)CCCCC2
Tekniske data
Kemisk navn eller materiale R-(+)-Venlafaxine
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C17H27NO2/c1-18(2)13-16(17(19)11-5-4-6-12-17)14-7-9-15(20-3)10-8-14/h7-10,16,19H,4-6,11-13H2,1-3H3/t16-/m0/s1
Formel vægt 277.2042
Procent renhed >95%