Få mere at vide
(R,S,S,S)-Nebivolol Hydrochloride, TRC
Artikelnummer. 30468779
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30468779 10 mg
30472723 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30468779 Leverandør Toronto Research Chemicals Leverandørnr. TRCN38790510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R,S,S,S)-Nebivolol Hydrochloride
CAS 920299-33-8
Molekylær formel C22H25F2NO4.ClH
Mængde 10 mg
Synonym Nebivolol Impurity A HCl, L-Nebivolol HCL, Nebivolol (RS,SS) HCl
InChI formel InChI=1S/C22H25F2NO4.ClH/c23-15-3-7-19-13(9-15)1-5-21(28-19)17(26)11-25-12-18(27)22-6-2-14-10-16(24)4-8-20(14)29-22;/h3-4,7-10,17-18,21-22,25-27H,1-2,5-6,11-12H2;1H/t17-,18-,21-,22+;/m0./s1
SMIL Cl.O[C@@H](CNC[C@H](O)[C@H]1CCc2cc(F)ccc2O1)[C@@H]3CCc4cc(F)ccc4O3
IUPAC navn (1S)-1-[(2S)-6-fluorochroman-2-yl]-2-[[(2S)-2-[(2R)-6-fluorochroman-2-yl]-2-hydroxy-ethyl]amino]ethanol;hydrochloride
Molekylvægt (g/mol) 441.9
Formel vægt 441.1518