Få mere at vide
(R,S)-Norcotinine, TRC
Artikelnummer. 17984705
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17984705 25 mg
17961285 10 mg
17942505 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17984705 Leverandør Toronto Research Chemicals Leverandørnr. TRCN66200025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17708-87-1
Molekylær formel C9 H10 N2 O
Molekylvægt (g/mol) 162.19
Synonym 2-Pyrrolidinone, 5-(3-pyridinyl)-, (±)-, Norcotinine, (±)- (8CI), (RS)-Norcotinine, (±)-Demethylcotinine, (±)-Norcotinine
IUPAC navn 5-pyridin-3-ylpyrrolidin-2-one
SMIL O=C1CCC(N1)c2cccnc2
Tekniske data
Kemisk navn eller materiale (RS)-Norcotinine
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C9H10N2O/c12-9-4-3-8(11-9)7-2-1-5-10-6-7/h1-2,5-6,8H,3-4H2,(H,11,12)
Formel vægt 162.0793
Procent renhed >95%