missing translation for 'onlineSavingsMsg'
Få mere at vide

(R,S)-N-Nitroso Anatabine-2,4,5,6-d4 (1 mg/ml i Methanol), TRC

Artikelnummer. 30395499
Ansicht ändern
Click to view available options
Mængde:
1x1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30395499 1x1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30395499 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCKIT18401X1ML
 Begrænset vare
Forventet forsendelse: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (R,S)-N-Nitroso Anatabine-2,4,5,6-d4
CAS 1020719-69-0
Molekylær formel C10 2H4 H7 N3 O
Mængde 1x1 mL
Synonym 2,3'-Bipyridine-2',4',5',6'-d4, 1,2,3,6-tetrahydro-1-nitroso-, (RS)-N-Nitrosoanatabine D4 (pyridinyl D4), (R,S)-N-Nitroso Anatabine-2,4,5,6-d4, (R,S)-N-Nitrosoanatabine D4 (pyridinyl D4), 1,2,3,6-Tetrahydro-1-nitroso-2,3'-bipyridine-2',4',5',6'-d4 (ACI)
InChI formel InChI=1S/C10H11N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h1-4,6,8,10H,5,7H2/i3D,4D,6D,8D
SMIL [2H]c1nc([2H])c(C2CC=CCN2N=O)c([2H])c1[2H]
IUPAC navn 2,3,4,6-tetradeuterio-5-(1-nitroso-3,6-dihydro-2H-pyridin-2-yl)pyridin
Molekylvægt (g/mol) 193.24
Formel vægt 193.1153
Koncentration eller sammensætning (efter analyt eller komponenter) 1 mg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.