Få mere at vide
(R,S)-Cotinine-d3 N-Oxide, TRC
Artikelnummer. 30368248
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30368248 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30368248 Leverandør Toronto Research Chemicals Leverandørnr. TRCC7252035MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R,S)-Cotinine N-Oxide-methyl-d3
CAS 1215561-37-7
Renhedsgrad noter HPLC
Molekylær formel C10 2H3 H9 N2 O2
Mængde 5 mg
InChI formel InChI=1S/C10H12N2O2/c1-11-9(4-5-10(11)13)8-3-2-6-12(14)7-8/h2-3,6-7,9H,4-5H2,1H3/i1D3
SMIL [2H]C([2H])([2H])N1C(CCC1=O)c2ccc[n+]([O-])c2
IUPAC navn 5-(1-oxidopyridin-1-ium-3-yl)-1-(trideuteriomethyl)pyrrolidin-2-one
Molekylvægt (g/mol) 195.23
Formel vægt 195.1087
Procent renhed >95