Få mere at vide
(R,S)-alpha-Methyldopamine Hydrobromide, Mikromol™
Artikelnummer. 30051739
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051739 25 mg
30049473 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051739 Leverandør Mikromol Leverandørnr. MM0384.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(R,S)-alpha-Methyldopamine Hydrobromide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R,S)-alpha-Methyldopamine Hydrobromide
CAS 3459-15-2
Analyt- eller komponentnavne (R,S)-alpha-Methyldopamine Hydrobromide
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C9 H13 N O2 . Br H
Certificeringer/compliance ISO 9001
Synonym (R,S)-alpha-Methyldopamine Hydrobromide
InChI formel InChI=1S/C9H13NO2.BrH/c1-6(10)4-7-2-3-8(11)9(12)5-7;/h2-3,5-6,11-12H,4,10H2,1H3;1H
SMIL Br.CC(N)Cc1ccc(O)c(O)c1
IUPAC navn 4-(2-aminopropyl)benzene-1,2-diol;hydrobromide
Molekylvægt (g/mol) 248.12
Mængde 25 mg
Formel vægt 247.0208 g/mol