Få mere at vide
(R,S)-1-Tosyl Glycerol-d5, TRC
Artikelnummer. 30453613
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30453613 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453613 Leverandør Toronto Research Chemicals Leverandørnr. TRCT66400250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R,S)-1-Tosyl Glycerol-d5
CAS 928623-32-9
Molekylær formel C10H9D5O5S
Mængde 50 mg
Synonym 1-Tosyloxy-2,3-propanediol-d5, 2,3-Dihydroxypropyl Tosylate-d5, p-Toluenesulfonic Acid Glyceryl Ester-d5, 1-p-Toluenesulfonate Glycerol-d5, 1,2,3-Propanetriol 1-(4-Methylbenzenesulfonate)-d5
InChI formel InChI=1S/C10H14O5S/c1-8-2-4-10(5-3-8)16(13,14)15-7-9(12)6-11/h2-5,9,11-12H,6-7H2,1H3/i6D2,7D2,9D
SMIL [2H]C([2H])(O)C([2H])(O)C([2H])([2H])OS(=O)(=O)c1ccc(C)cc1
IUPAC navn (1,1,2,3,3-pentadeuterio-2,3-dihydroxypropyl) 4-methylbenzenesulfonate
Molekylvægt (g/mol) 251.31
Formel vægt 251.0876