Få mere at vide
(R,S)-1-Methyl-3-nicotinoylpyrrolidone-d4, TRC
Artikelnummer. 30465819
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30465819 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465819 Leverandør Toronto Research Chemicals Leverandørnr. TRCM323433100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R,S)-1-Methyl-3-nicotinoylpyrrolidone-d4
CAS 764661-22-5
Molekylær formel C11H8D4N2O2
Mængde 100 mg
Synonym (+/-)-1-Methyl-3-(3-pyridinylcarbonyl)-2-pyrrolidinone-d4
InChI formel InChI=1S/C11H12N2O2/c1-13-6-4-9(11(13)15)10(14)8-3-2-5-12-7-8/h2-3,5,7,9H,4,6H2,1H3/i2D,3D,5D,7D
SMIL [2H]c1nc([2H])c(C(=O)C2CCN(C)C2=O)c([2H])c1[2H]
IUPAC navn 1-methyl-3-(2,4,5,6-tetradeuteriopyridine-3-carbonyl)pyrrolidin-2-one
Molekylvægt (g/mol) 208.25
Formel vægt 208.115