Få mere at vide
(R)-RuCl[(p-cymene)(BINAP)]Cl, TRC
Artikelnummer. 30421738
Skift visning
Mængde:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421738 25 mg
30409043 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30421738 Leverandør Toronto Research Chemicals Leverandørnr. TRCR70157625MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-RuCl[(p-cymene)(BINAP)]Cl
CAS 145926-28-9
Molekylær formel C45 H35 Cl P2 Ru . C10 H14 . Cl
Mængde 25 mg
Synonym Chloro[(R)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride
InChI formel InChI=1S/C44H32P2.C10H14.CH3.2ClH.Ru/c1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38;1-8(2)10-6-4-9(3)5-7-10;;;;/h1-32H;4-8H,1-3H3;1H3;2*1H;/q;;;;;+2/p-2
SMIL [Cl-].CC(C)c1ccc(C)cc1.C[Ru+]2(Cl)[P](c3ccccc3)(c4ccccc4)c5ccc6ccccc6c5c7c(ccc8ccccc78)[P]2(c9ccccc9)c%10ccccc%10
Molekylvægt (g/mol) 943.901
Formel vægt 943.173