Learn More
(R)-rel-2,5,7,8-Tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-yl acetate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 52225-20-4 |
| Molecular Formula | C31H52O3 |
| MDL Number | MFCD00006847 |
| Quantity | 25 g |
| SMILES | CC(C)CCCC(C)CCCC(C)CCCC1(C)CCC2=C(O1)C(C)=C(C)C(OC(C)=O)=C2C |
| Molecular Weight (g/mol) | 472.74 |
| Percent Purity | 98% |
| Chemical Name or Material | (R)-rel-2,5,7,8-Tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-yl acetate |