Få mere at vide
(R)-Rabeprazole-d3 Sodium Salt, TRC
Artikelnummer. 30419479
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30419479 10 mg
30408976 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419479 Leverandør Toronto Research Chemicals Leverandørnr. TRCR07052210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1216494-11-9
Molekylær formel C18H17D3N3NaO3S
Molekylvægt (g/mol) 384.44
Synonym 2-[(R)-[[4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl]methyl]sulfinyl]-1H-benzimidazole-d3 Sodium Salt, R-(+)-Rabeprazole-d3 Sodium Salt
IUPAC navn sodium (R)-2-(((4-(3-(methoxy-d3)propoxy)-3-methylpyridin-2-yl)methyl)sulfinyl)benzo[d]imidazol-1-ide
SMIL O=[S@](CC1=NC=CC(OCCCOC([2H])([2H])[2H])=C1C)C2=NC3=CC=CC=C3N2[Na]
Tekniske data
Kemisk navn eller materiale (S)-Rabeprazole-d3 Sodium Salt
Mængde 10 mg
InChI formel InChI=1S/C18H20N3O3S.Na/c1-13-16(19-9-8-17(13)24-11-5-10-23-2)12-25(22)18-20-14-6-3-4-7-15(14)21-18;/h3-4,6-9H,5,10-12H2,1-2H3;/q-1;+1/t25-;/m1./s1/i2D3;
Formel vægt 384.13