Få mere at vide
[R-(R*,R*)]-(1,2-Dihydroxypropyl)phosphonic Acid, TRC
Artikelnummer. 30135733
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135733 100 mg
30010499 50 mg
30008608 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135733 Leverandør Toronto Research Chemicals Leverandørnr. TRCD454455100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 132125-60-1
Molekylær formel C3 H9 O5 P
Molekylvægt (g/mol) 156.07
Synonym Phosphonic acid, (1,2-dihydroxypropyl)-, [R-(R*,R*)]- (9CI)
IUPAC navn [(1R,2R)-1,2-dihydroxypropyl]phosphonic acid
SMIL C[C@@H](O)[C@H](O)P(=O)(O)O
Tekniske data
Kemisk navn eller materiale [R-(R*,R*)]-(1,2-Dihydroxypropyl)phosphonic Acid
Mængde 100 mg
InChI formel InChI=1S/C3H9O5P/c1-2(4)3(5)9(6,7)8/h2-5H,1H3,(H2,6,7,8)/t2-,3-/m1/s1
Formel vægt 156.0188